missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Accessories for Cole-Parmer™ MP-250D Series Digital Melting Point Apparatus

Accessories compatible with digital melting point apparatus.

275.00 € - 969.00 €

Specifications

For Use With (Equipment) MP-250D Series Digital Melting Point Apparatus
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11807493
View Documents
Cole-Parmer™
03010-69
Serial Printer
969.00 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
 
13484879
View Documents
Cole-Parmer™
03010-65
Calibration Key
275.00 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
 
Description

Description

  • 1 year warranty.
Specifications

Specifications

MP-250D Series Digital Melting Point Apparatus
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Accessories for Cole-Parmer™ MP-250D Series Digital Melting Point Apparatus

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.