missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Accessories for Peristaltic Pump Systems

Upgrade pumping and dispensing capabilities with Accessories for Masterflex™ Peristaltic Pump Systems.

Brand:  Masterflex™ 07523-95

 View more versions of this product

Product Code. 15497144

  • 254.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Upgrade pumping and dispensing capabilities with Accessories for Masterflex™ Peristaltic Pump Systems.

  • Accessory items for the Masterflex™ Peristaltic Pump Systems

Services

2 Years

Specifications

Specifications

ISO 9002, CE
Remote I/O
Masterflex drives with DB-25 connection and remote control capability
25 ft.
External Control-Input: Current, voltage, start/stop, direction, prime; External Control-Output: Speed (current), speed (voltage), motor running, tachometer, local/remote, alarm, auxiliary 24 V
1.26 lb.
Gray
Fluid handling
Masterflex Cable Assembly for Remote Control, 25 ft. (7.6 m) cable with stripped-wire ends, DB25(M) connector
7.6 m
Analog
571.53 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.