missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Alumina Notched Plate

70.69 € - 434.00 €

Specifications

Type Notched Alumina Plate
Material Alumina
Length (Metric) 10 cm
Width (Metric) 8 cm
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11735393
View Documents
Cytiva
SE202N-10
SE 250 Mini Vertical Gel Electrophoresis Unit
434.00 €
Pack of 10
In Stock
to see stock.
Add to basket
 
10279714
View Documents
Cytiva
SE202N
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
70.69 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Notched Alumina Plate
Alumina
10 cm
8 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.