missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Accessories for SE 660 Vertical Electrophoresis Unit

93.88 € - 186.00 €

Specifications

For Use With (Equipment) SE 660 Vertical Electrophoresis Unit
Type Glass Plate
Material Glass
Quantity 2/Pk.
Length (Metric) 18 cm
View More Specs

Products 2
Product Code Brand Type Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Type Length (Metric) Width (Metric) Price Quantity & Availability  
10713267
View Documents
Cytiva
SE6402
Glass Plate 18 cm 8 cm
186.00 €
Pack of 2
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
11520764
View Documents
Cytiva
SE6102
Glass Plate 18 cm 16 cm
93.88 €
Pack of 2
In Stock
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

SE 660 Vertical Electrophoresis Unit
Glass
18 cm
Glass Plate
2/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.