missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ R 1389 Propeller Stirrer
With a flow-efficient design for drawing the material to be mixed from the top and the bottom.
Brand: IKA™ 0002343600
650.75 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- With 3 blades
- Minimum shearing forces
- Used at medium to high speeds
- PTFE-coated
- Stirrer diameter of 75mm, shaft diameter of 8mm, shaft length of 350mm

Specifications
75 mm | |
A wide range of IKA™ stirrers | |
800 rpm Max |
8 mm | |
350 mm shaft | |
3-bladed Propeller Stirrer |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKA™ R 1389 Propeller Stirrer > R 1389, Stirrer, Propeller
Spot an opportunity for improvement?Share a Content Correction