Learn More
Thermo Scientific™ Acclaim™ PepMap™ 100 C18 HPLC Columns
Use for high-resolution tryptic, natural, and synthetic peptide analyses. Its high loading capacity is exceptional for low abundant peptides in complex proteomic samples.
287.64 € - 1489.01 €
Specifications
Pore Size | 100 Å |
---|---|
USP Type | L1 |
Stationary Phase | C18 |
Phase | Reversed Phase |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Type | Price | Quantity & Availability | ||||||
11342263
|
Thermo Scientific™
164567 |
50 mm | 0.075 mm | 3 μm | Analytical Column, NanoViper | N/A | N/A | N/A | |||||
11352263
|
Thermo Scientific™
164568 |
150 mm | 0.075 mm | 3 μm | Analytical Column |
1189.41 €
Each |
In Stock
Log in to see stock. |
||||||
11362263
|
Thermo Scientific™
164569 |
250 mm | 0.075 mm | 3 μm | Analytical Column |
1298.71 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
11372263
|
Thermo Scientific™
164570 |
500 mm | 0.075 mm | 3 μm | Analytical Column |
1489.01 €
Each |
In Stock
Log in to see stock. |
||||||
11302273
|
Thermo Scientific™
164572 |
150 mm | 1 mm | 3 μm | Analytical Column |
889.81 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
11312113
|
Thermo Scientific™
160434 |
5 mm | 1 mm | 5 μm | Trap Cartridge |
559.34 €
Each |
In Stock
Log in to see stock. |
||||||
11362113
|
Thermo Scientific™
160454 |
5 mm | 0.3 mm | 5 μm | Trap Cartridge | N/A | N/A | N/A | |||||
11322113
|
Thermo Scientific™
160438 |
15 mm | 1 mm | 5 μm | Trap Cartridge | N/A | N/A | N/A | |||||
11382243
|
Thermo Scientific™
164197 |
150 mm | 0.1 mm | 5 μm | Trap Column | N/A | N/A | N/A | |||||
11332253
|
Thermo Scientific™
164213 |
150 mm | 0.2 mm | 5 μm | Trap Column |
328.42 €
Pack of 2 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
11392243
|
Thermo Scientific™
164199 |
150 mm | 0.1 mm | 5 μm | Trap Column |
402.48 €
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11352103
|
Thermo Scientific™
160323 |
150 mm | 0.075 mm | 5 μm | Analytical Column | N/A | N/A | N/A | |||||
11362103
|
Thermo Scientific™
160326 |
250 mm | 0.075 mm | 5 μm | Analytical Column | N/A | N/A | N/A | |||||
11352023
|
Thermo Scientific™
164562 |
150 mm | 0.05 mm | 2 μm | Analytical Column, NanoViper | N/A | N/A | N/A | |||||
11372013
![]() |
Thermo Scientific™
164537 |
150 mm | 0.3 mm | 2 μm | Analytical Column |
880.81 €
Each |
In Stock
Log in to see stock. |
||||||
11392253
|
Thermo Scientific™
164563 |
50 mm | 0.75 mm | 2 μm | Analytical Column, NanoViper | N/A | N/A | N/A | |||||
11382013
|
Thermo Scientific™
164540 |
500 mm | 0.75 mm | 2 μm | Analytical Column | N/A | N/A | N/A | |||||
11342013
![]() |
Thermo Scientific™
164534 |
150 mm | 0.075 mm | 2 μm | Analytical Column |
1150.73 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
11342023
|
Thermo Scientific™
164561 |
50 mm | 0.05 mm | 2 μm | Analytical Column, NanoViper | N/A | N/A | N/A | |||||
11332023
|
Thermo Scientific™
164560 |
50 mm | 0.3 mm | 2 μm | Analytical Column |
898.28 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
15597520
|
Thermo Scientific™
164941 |
250 mm | 0.75 mm | 2 μm | Analytical Column |
1272.57 €
Each |
In Stock
Log in to see stock. |
||||||
11352013
|
Thermo Scientific™
164535 |
150 mm | 0.075 mm | 3 μm | Trap Column |
623.63 €
Pack of 2 |
In Stock
Log in to see stock. |
||||||
15810705
|
Thermo Scientific™
164738 |
500 mm | 0.075 mm | 2 μm | Analytical Column | N/A | N/A | N/A | |||||
15820705
|
Thermo Scientific™
164739 |
500 mm | 0.075 mm | 3 μm | Analytical Column | N/A | N/A | N/A | |||||
15567530
|
Thermo Scientific™
164942 |
500 mm | 0.075 mm | 2 μm | Analytical Column |
1460.68 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
15572920
|
Thermo Scientific™
164939 |
750 mm | 0.075 mm | 2 μm | Analytical nano Column | N/A | N/A | N/A | |||||
15800705
|
Thermo Scientific™
164454 |
50 mm | 1 mm | 2 μm | Analytical Column, NanoViper |
1431.16 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15588090
|
Thermo Scientific™
164946 |
20 mm | 75 μm | 3 μm | Trap Column |
611.04 €
Each |
In Stock
Log in to see stock. |
||||||
12739835
|
Thermo Scientific™
164704 |
150 mm | 0.05 mm | 2 μm | Analytical Column | N/A | N/A | N/A | |||||
15527660
|
Thermo Scientific™
164943 |
150 mm | 0.05 mm | 2 μm | Analytical Column |
1116.02 €
Each |
In Stock
Log in to see stock. |
||||||
11382253
|
Thermo Scientific™
164261 |
250 mm | 0.075 mm | 3 μm | Analytical Column | N/A | N/A | N/A | |||||
11342103
|
Thermo Scientific™
160321 |
150 mm | 0.075 mm | 3 μm | Analytical Column | N/A | N/A | N/A | |||||
11312263
|
Thermo Scientific™
164564 |
150 mm | 0.1 mm | 5 μm | Trap Column | N/A | N/A | N/A | |||||
15517460
|
Thermo Scientific™
164940 |
150 mm | 0.075 mm | 2 μm | Analytical Column |
1094.57 €
Each |
In Stock
Log in to see stock. |
||||||
17294434
|
Thermo Scientific™
164750 |
150 mm | 0.1 mm | 5 μm | Trap Column |
623.63 €
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||||
17204444
|
Thermo Scientific™
164751 |
150 mm | 0.3 mm | 3 μm | Analytical Column, Nanoviper | N/A | N/A | N/A | |||||
13486836
|
Thermo Scientific™
164711 |
150 mm | 1 mm | 2 μm | Analytical Column |
855.95 €
Each |
In Stock
Log in to see stock. |
||||||
15518080
|
Thermo Scientific™
164944 |
250 mm | 0.05 mm | 2 μm | Column |
1272.57 €
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15588080
|
Thermo Scientific™
164945 |
500 mm | 0.05 mm | 2 μm | Column |
1460.57 €
Each |
In Stock
Log in to see stock. |
||||||
Description
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ PepMap™ 100 C18 LC Columns. PepMap LC columns offer lengths targeting fast or high resolution peptide separation.
- High resolution in protein identification, biomarker discovery, and systems biology
- Particular loadability allows highest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
- Preassembled with nanoViper column inlet fittings for easy column installation
Key advantages of nanoViper fittings
- Ease of use, no sleeve for small capillaries
- Tool-free assembly
- No column damage from overtightening: capable of withstanding pressures up to 1000 bars and is compatible with third-party valves and unions
- No experimental failure due to bad connections
Ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides as well as LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.
- High loading capacity
- Use for the analysis of low abundant peptides often found in complex proteomics samples
- Select the column length and I.D. for your throughput, sensitivity, and amount of sample available requirements
- Choose from 50, 150, 250, 500mm lengths and 50, 75, 300 and 1000μm I.D.
- Superior protein and peptide separations
- Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
- Outstanding separation efficiency
- Superior resolution and recovery
- Easy installation with nanoViper™ Fingertight Fittings
PepMap nanocolumns are also available pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fitting for a dead-volume-free connection to 1000 bar.
- Requires no tools
- Provides virtually zero-dead-volume connections
- Withstands pressures up to a 1000 bar
About Nano, capillary, and micro Acclaim PepMap columns
Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and microcolumns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and microcolumn in a variety of chemistries for peptides and protein analysis.
Use PepMap Trap columns for desalting and preconcentration of peptides before LC separation with MS detection. The trap columns provide the highest efficiencies for one-dimensional peptide mapping experiments and 2D-LC analyses.
Specifications
100 Å | |
L1 | |
C18 | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.