missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ AmG C18 HPLC Columns

Meet today’s demands for aminoglycoside antibiotic analyses, including drug purity characterization & quantification. They have unique tolerance to harsh conditions.

Brand:  Thermo Scientific™ 088757

986.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15522930

  • 991.36 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get a high-performance, reversed-phase columns for aminoglycoside analysis. The robust and rugged Thermo Scientific™ Acclaim™ AmG C18 HPLC column provides exceptional tolerance required for aminoglycosides ion-pairing chromatographic separations. These silica-based C18 columns are optimized for various aminoglycoside antibiotic analyses, including drug purity characterization and quantification, therapeutic drug monitoring, and residual control testing in different matrices. These columns are stable at low pH, high temperature, organic solvents, and aqueous mobile phases.

Highly Stable for Aminoglycoside Analyses

The Acclaim AmG C18 column is specifically designed for ion-pairing reversed-phase HPLC (IP-RPLC) analysis of aminoglycoside antibiotics using volatile perfluorinated carboxylic acids as the ion-pairing reagent, such as 100 mM trifluoroacetic acid (TFA). The Acclaim AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions. This specialty column exhibits excellent selectivity and high resolution for the aminoglycoside antibiotics analysis. In addition, it is easy to use because simple aqueous mobile phase (e.g. 100 mM TFA) without organic solvent and pH adjusting buffer is sufficient in most cases.

  • Excellent selectivity for aminoglycosides
  • Superior tolerance towards acidic conditions
  • High efficiency and throughput
  • Ease of use

The column can be used with UV, CAD, ELSD detectors

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
150 mm
3 μm
120 Å
<80°C
C18
Spherical, High Purity Silica
Reversed Phase
4.6 mm
8000 psi (550 bar)
0.5 to 10
Each
L1
Acclaim
Aminoglycoside Analysis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.