Learn More
Thermo Scientific™ Acclaim™ Carbonyl C18 HPLC Columns
Use for superior resolution and high-throughput separation of carbonyls, dinitrophenylhydrazine (DNPH) derivatives of aldehydes and ketones.
244.84 € - 1514.65 €
Specifications
pH | 2.5 to 7.5 |
---|---|
Pore Size | 120 Å |
Surface Area | 300 m²/g |
Temperature | 50°C |
Endcapped | Yes |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
12366543
|
Thermo Scientific™
077972 |
100 mm | 2.1 mm | 2.2 μm |
950.39 €
Each |
In Stock
Log in to see stock. |
||||
12396543
|
Thermo Scientific™
077973 |
150 mm | 2.1 mm | 2.2 μm |
983.29 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
12366553
|
Thermo Scientific™
077974 |
100 mm | 3 mm | 2.2 μm |
988.56 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
15237468
|
Thermo Scientific™
079009 |
250 mm | 3 mm | 3 μm |
1514.65 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||
15247468
|
Thermo Scientific™
079010 |
150 mm | 3 mm | 3 μm |
964.86 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
15257468
|
Thermo Scientific™
079011 |
150 mm | 2.1 mm | 3 μm |
1059.07 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||
15123707
|
Thermo Scientific™
079012 |
10 mm | 2.1 mm | 5 μm |
244.84 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||
15267468
|
Thermo Scientific™
079013 |
10 mm | 3 mm | 5 μm |
268.91 €
Each |
In Stock
Log in to see stock. |
||||
15277468
|
Thermo Scientific™
079014 |
10 mm | 4.6 mm | 5 μm |
256.69 €
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||
Description
Use the Thermo Scientific™ Acclaim™ Carbonyl C18 LC Columns for superior resolution and high-throughput separation of dinitrophenylhydrazine (DNPH) derivatives of aldehydes and ketones. This silica-based, reversed-phase column provides ideal selectivity and baseline resolution of DNPH derivatives regulated by various official methods, including U.S. EPA 554, EPA 8315, EPA 1667, EPA TO-11, and CARB 1004.
Superior Performance for Challenging Aldehyde and Ketone Analysis
- High efficiency separations for excellent UHPLC performance
- Rugged columns with good lot-to-lot reproducibility
- Proven robust methods
Wide Variety of Column Dimensions to Meet Your Analysis Needs
- 2.2, 3 and 5μm particle sizes; smaller particle sizes provide high peak efficiency for fast analysis
- 2.1, 3.0 and 4.6mm diameters; small diameter columns (2.1 and 3.0mm) consume less solvents and produce less chemical waste
- 100, 150 and 250mm lengths
For high-throughput applications, use the 2.1×100mm column format combined with high linear velocity. When high resolution power is needed, use a longer column (2.1×150mm) at an optimal linear velocity to achieve even better resolution.
About the Family of Acclaim Specialty HPLC Columns
Acclaim specialty columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease of use.
- Use-tested to guarantee consistent separations
- Ideal selectivity for separating specific target analytes
- Excellent peak shapes
- Performance tested to ensure high quality and reliability for specific chromatographic separations
Specifications
2.5 to 7.5 | |
300 m²/g | |
Yes |
120 Å | |
50°C | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.