missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ Trinity P1 HPLC Columns

Use these mixed-mode columns for simultaneous separation of pharmaceutical drug substances and their counter ions with high efficiency.

Brand:  Thermo Scientific™ 075564

 View more versions of this product

Product Code. 11372073

  • 1120.04 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Simultaneously separate pharmaceutical drug substances and their counterions with high-efficiency Thermo Scientific™ Acclaim™ Trinity P1 LC Columns. A unique trimodal surface chemistry provides concurrent reversed-phase, cation-exchange and anion-exchange functionalities for maximum flexibility in method development. Easily optimize selectivity by adjusting mobile phase buffer concentration, pH and solvent content—either jointly or independently—to accelerate chromatographic separations and increase productivity.

The Acclaim Trinity P1 stationary phase, based on Nanopolymer Silica Hybrid (NSH) technology, consists of high-purity porous, spherical 3μm silica particles, coated with charged nanopolymer beads. The unique surface chemistry includes an inner-pore area modified with an organic layer that provides both reversed-phase and anion-exchange properties. The outer-pore surface, conversely, is modified with cation-exchange functionality for unparalleled chromatographic performance. 
 
Superior Performance for Pharmaceutical Applications
  • Ideal selectivity for simultaneous separation of Active Pharmaceutical Ingredients (API) and counterions (basic, neutral and acidic analytes)
  • Effective retention of hydrophilic ionic and ionizable analytes without ion-pairing reagents 
  • Distinctive spatial separation of the anion-exchange and cation-exchange regions
  • Flexible method development with each retention mechanism controlled independently
  • Low column bleed; compatible with mass spectrometry (MS), charged aerosol detection (CAD) and evaporative light scattering detectors (ELSD) 
  • Available in 2.1 and 3.0mm diameters and 50, 100 and 150mm lengths
Complement the Acclaim Trinity P1 column with the Acclaim Trinity P2 column to provide a total solution for pharmaceutical counterion analysis by HPLC. Use the Acclaim Trinity P2 column to separate counterions, including monovalent and divalent cations or anions.
 
About the Family of Acclaim Specialty HPLC Columns
Acclaim specialty columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease of use.
  • Use tested to guarantee consistent separations
  • Ideal selectivity for separating specific target analytes
  • Excellent peak shapes 
  • Performance tested to ensure high quality and reliability for specific chromatographic separations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
ELSD, UV Detector
4500 psi (310 bar)
2.5 to 7
Each
45°C
Acclaim
Mixed Mode
Reversed Phase and Ion Exchange
2.1 mm
150 mm
3 μm
300 Å
100 m²/g
Reversed Phase/Anion Exchange/Cation Exchange
Spherical, Nanopolymer Hybrid Silica
Proprietary
Analytical Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.