missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Accu20 Ultrapure Water System
Fisherbrand™ Accu20 Ultrapure Water System used to produce Type I ultrapure water and RO water on demand from tap water directly.
5835.00 € - 7105.00 €
Specifications
Conductivity | RO water at 25°C: Typically <20μS/cm |
---|---|
Flow Rate | RO water (at 25°C ): >0.5L/min.; Ultrapure water ( Type I ) (at 25°C ): >0.5L/min. |
Resistivity | 18.2 Mohm.cm |
Feedwater Pressure | 15 to 90 psi (1 to 6 bar ) |
Model | Accu20 |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15489689
|
Fisherbrand™
RD0A0400EK |
ACCU20 Ultrapure Water System |
5835.00 €
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15499689
|
Fisherbrand™
RD0A040UEK |
ACCU20 UV Ultrapure Water System |
7105.00 €
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
Description
- Compact system provides easy way to produce ultrapure water or RO water on demand from tap water directly with no additional pre-treatment filters
- Equipped with safeguard AccuDuo P pack cartridge and dual-column AccuDuo U Pack ultrapurification cartridge to ensure optimum quality of final product water
- No external storage tank required, which eliminate risk of water degration by contaminants from container or tubing
- Fully automatic controlled and maintained system contains automatic cleaning cycles and selfmaintenance functions for RO membrane, which increases lifetime and efficiency of RO membranes as well as cartridges
- Automatic hourly system recirculation in loop prevents bacterial growth and maintain water purity at all times
- System status and key quality parameters are clearly displayed on large LCD screen for easy system performance monitoring
Applications:
- HPLC mobile phase preparation
- Preparation of reagent blank solutions
- As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
- Preparation of buffers and culture media for mammalian cell culture
- Preparation of molecular biology reagents, etc.
Certifications
Meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards
Specifications
RO water at 25°C: Typically <20μS/cm | |
18.2 Mohm.cm | |
Accu20 | |
76.2 in. | |
18.8 in. | |
20 in. |
RO water (at 25°C ): >0.5L/min.; Ultrapure water ( Type I ) (at 25°C ): >0.5L/min. | |
15 to 90 psi (1 to 6 bar ) | |
5°C to 45°C | |
30 cm | |
48 cm | |
51 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title