missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Accu500 Water System

Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.

8265.00 € - 11795.00 €

Specifications

Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Water Type Tap Water
Resistivity 18.2 Mohm.cm
Feedwater Pressure 1 to 6 bar
Model Accu500
View More Specs

Products 12
Product Code Brand Type RO Flow Rate Price Quantity & Availability  
Product Code Brand Type RO Flow Rate Price Quantity & Availability  
15409699
View Documents
Fisher Scientific™
RD0P0100EK
Accu500 Ultrapure Water System 10 L/hr.
8265.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15419699
View Documents
Fisher Scientific™
RD0P010UEK
Accu500 UV Ultrapure Water System 10 L/hr.
9245.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15409709
View Documents
Fisher Scientific™
RD0PP10UEK
Accu500 UV Ultrapure Water System, with Dispenser 10 L/hr.
10825.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15429699
View Documents
Fisher Scientific™
RD0P0150EK
Accu500 Ultrapure Water System 15 L/hr.
8420.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15439699
View Documents
Fisher Scientific™
RD0P015UEK
Accu500 UV Ultrapure Water System 15 L/min.
9395.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15419709
View Documents
Fisher Scientific™
RD0PP15UEK
Accu500 UV Ultrapure Water System, with Dispenser 15 L/hr.
10985.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15449699
View Documents
Fisher Scientific™
RD0P0200EK
Accu500 Ultrapure Water System 20 L/hr.
8915.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15459699
View Documents
Fisher Scientific™
RD0P020UEK
Accu500 UV Ultrapure Water System 20 L/hr.
9895.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15429709
View Documents
Fisher Scientific™
RD0PP20UEK
Accu500 UV Ultrapure Water System, with Dispenser 20 L/hr.
11475.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15469699
View Documents
Fisher Scientific™
RD0P0300EK
Accu500 Ultrapure Water System 30 L/hr.
9235.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15439709
View Documents
Fisher Scientific™
RD0PP30UEK
Accu500 UV Ultrapure Water System, with Dispenser 30 L/hr.
11795.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15479699
View Documents
Fisher Scientific™
RD0P030UEK
Accu500 UV Ultrapure Water System 30 L/hr.
10210.00 €
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
Description

Description

  • The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility

Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:

Ultrapure water

  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.

RO water

  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving
Specifications

Specifications

<0.001 EU/mL with a terminal ultrafiltration cartridge
18.2 Mohm.cm
Accu500
1.5 L/min.
30 cm
51 cm
Tap Water
1 to 6 bar
<1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
5°C to 45°C
48 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.