Learn More
Fisherbrand™ AccuDuo Cartridge
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
Brand:  Fisherbrand™ RR600Q2KT
 View more versions of this product
Product Code. 15499729
  • 544.00 € / Each
Estimated Shipment: 18-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
Specifications
Accu20, Accu500
U Pack with a 0.2μm PES Capsule Filter
Ultrapure water for critical laboratory applications
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ AccuDuo Cartridge >