Learn More
Fisherbrand™ accumet™ AB150 pH Benchtop Meters
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
710.00 € - 1065.00 €
Specifications
Accuracy (mV) | ±0.2 |
---|---|
Accuracy (pH) | ±0.002 |
Accuracy (Temperature) | ±0.3°C |
Data Management Details | Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format |
Outputs | RS-232 (phono plug), mini-B USB, stirrer |
Product Code | Brand | Model | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
12880633
|
Fisherbrand™
01X605801 |
AB150 pH BioBasic Kit | Meter, TRIS Compatible accuTupH pH Electrode (13-620-183A), ATC Probe (13-620-19), Electrode Arm, 110/220V Power Supply | Without NIST-traceable pH Certificate | N/A | ||||
12870633
|
Fisherbrand™
01X605701 |
AB150 pH Kit | Meter, TRIS Compatible pH/ATC Electrode (13-620-631), Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply | Without NIST-traceable pH Certificate | N/A | ||||
12840633
|
Fisherbrand™
01X605601 |
AB150 pH Meter Only | Meter, Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply | Without NIST-traceable pH Certificate | N/A | ||||
Description
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
- Graphic backlit display with date and time, stability indicator and alarms
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with four pH buffer sets or custom buffer option
- pH slope and offset display
- One point relative mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation, displayed in °C or °F
- Password protection of calibration and setup modes
- 500 data set storage
- RS232 and USB interfaces
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Optional stirring probe (13-620-BSP) with five speed settings
NIST-Traceable Calibration Models (-CERT)
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory; includes NIST-Traceable certificate
Specifications
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Temperature Range: 0° to 100°C (32° to 212.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.3°C (±0.5°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Self-Test: Yes
- Inputs: BNC, mini-phone ATC, ground
- Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Includes:
13-636-AB150A: accumet AB150 pH meter, electrode stand, RS232 and USB cables, 100/240V universal power supply.
13-636-AB150: accumet AB150 pH meter, TRIS-compatible double junction refillable epoxy-body pH/ATC electrode (13-620-631), electrode stand, RS232 and USB cables, 100/240V universal power supply.
13-636-AB150B: accumet AB150 pH meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC temp probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply.
AB150ACERT: accumet AB150 pH meter, electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
AB150CERT: accumet AB150 pH meter, TRIS-compatible double junction refillable epoxy-body pH/ATC Electrode (13-620-631), electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
AB150BCERT: accumet AB150 pH meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC temp probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
Certifications
Select models available with Certificate of NIST Traceability
Warranty
3 years on meter
Specifications
±0.2 | |
±0.3°C | |
RS-232 (phono plug), mini-B USB, stirrer | |
0.001, 0.01, 0.1 | |
±2000 | |
0.1°C | |
500 | |
3 Years | |
50/60 Hz |
±0.002 | |
Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format | |
-2 to 20 | |
Up to 6 pre-set or 5 custom | |
0.1 mV | |
LCD | |
BNC, ATC | |
100/240 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.