missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ AE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price
Brand: Fisherbrand™ 13-636-AE151
Includes:
- Meter
- Single Junction Gel pH Electrode (#13-620-108B)
- ATC Probe (#13-620-22)
- Electrode Stand
- Literature
- Universal Power Adapter (90-240V, 50/60 Hz)
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Services
3 years on meter
Specifications
Meter, temperature probe and stand | |
±0.2 ±199.9 ±2 beyond ±200 | |
0 to 14 | |
±1999 | |
USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45 | |
0.1°C | |
Offset in 0.1 °C increments; Offset range: ±5 °C | |
BNC, 2.5 mm phono plug, reference pin tip, ground | |
AC/DC 9 V adapter (110/220 VAC, 50-60 Hz) |
±0.3°C | |
Recorder (±2000 mV) | |
0.01 | |
0.1 mV (±199.9 mV); 1 mV beyond ±200 mV | |
0.0°C to 100.0°C | |
Automatic or manual (0 to 100 °C) | |
100 data sets | |
Education Set |