missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AE150 Benchtop pH Meter

Product Code. 15534693 Shop All Fisherbrand Products
Click to view available options
Type:
3-in-1 Set
Bio Set
Education Set
Standard Set
Unit Size:
Each
This item is not returnable. Se returpolicy

Produktkod. 15534693

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Innehåller

Double Junction pH Electrode, Temperature Probe and Stand

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory

Garanti

3 years on meter

TRUSTED_SUSTAINABILITY

Specifikationer

Includes Meter, Double Junction pH Electrode, Temperature Probe and Stand
Accuracy (mV) ±0.2 ±199.9 ±2 beyond ±200
Outputs Recorder (±2000 mV)
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV
Calibration Points USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45
Temperature Range (Metric) 0.0°C to 100.0°C
Resolution (Temperature) 0.1°C
Temperature Probe Calibration Offset in 0.1 °C increments; Offset range: ±5 °C
Memory 100 data sets
Inputs BNC, 2.5 mm phono plug, reference pin tip, ground
Warranty 3 Year Meter Warranty
Temperature Compensation Automatic or manual (0 to 100 °C)
Type Bio Set
Electrical Requirements AC/DC 9 V adapter (110/220 VAC, 50-60 Hz)
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.