missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AET15 pH Tester

Large display screen - dual display LCD for enhanced readability, provides comprehensive meter information

Brand:  Fisherbrand™ 13-636-AET15

161.42 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


Code produit. 15553653

  • 194.00 â‚¬ / Each
In Stock
to see stock.
Edge
Découvrez plus d'offres spéciales
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Large display screen - dual display LCD for enhanced readability, provides comprehensive meter information

  • Waterproof, dustproof housing - IP67 rated, maintains the integrity of your tester even in harsh conditions. Plus it floats!
  • User-replaceable sensor - reuse the same meter body many times
  • Double junction sensor offers longer lasting usage
  • Push-button calibration with auto-buffer recognition - quick, easy calibration with no calibration mistakes
  • Automatic Temperature Compensation (ATC) - ensures optimum accuracy under fluctuating temperatures
  • Choice of USA or NIST pH buffer standards with up to 3 points
  • Temperature display readout simultaneously
  • Auto power-off saves battery power after non-use
  • “Hold” function freezes reading momentarily for easy viewing
  • Self-diagnostic for easy troubleshooting (e.g. battery indicator, message codes)
  • Enhanced power saving allows extended power consumption up to 500 hours with single battery set
Special functions:
  • Large dual custom display
  • Self-diagnostic
  • Hold function
  • ATC: yes
TRUSTED_SUSTAINABILITY
Spécification

Spécification

pH Tester Fisherbrand accumet AE15
±0.01
-1 to 15
USA: pH 4.01, 7.00, 10.01; NIST: pH 4.01, 6.86, 9.18
LCD, Dual Display with Battery Indicator, Error Message/Codes
4 x 1.5V alkaline button cell batteries
0.5°C/0.9°F
  • General: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required. accumet AET15 pH Tester meets requirements for many standard environmental and A
0.01
0.1°C, 0.1°F
Battery; auto power-off after 8.5 minutes
>500 hr.
Suggestions de produits

Suggestions de produits

Vidéos

Videos

  • Fisherbrand - accumet AET15 pH and AET30 conductivity testers
    pH or conductivity measurement has never been easier with the accumet AET pocket testers. Accurate, handy and user friendly, these are the best pocket testers in their class.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand accumet AET15 pH and AET30 conductivity testers

    FDS
    Documents Certificats
    Voir les accessoires
    Promotions

    Promotions

    Correction du contenu d'un produit

    Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

    Nom du produit
    Fisherbrand™ accumet™ AET15 pH Tester >

    En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

    Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis