missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ FE150 Benchtop pH Meter

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price.

Brand:  Fisherbrand™ S35922

333.49 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 17721739

  • 352.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet FE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Meter, Electrode Stand, Universal Power Adapter (90 to 240 V, 50/60 Hz)
±0.2 ±199.9 ±2 Beyond ±200
Recorder (±2000 mV)
0 to 14
0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV
0°C to 100°C
Automatic or Manual (0°C to 100°C)
100 Data Sets
3 Year Meter Warranty
Standard Set
±0.3°C
±0.01
USA: pH 1.68, 4.01, 7.00, 10.01, 12.45
±1999 mV
1 to 5 with Automatic Buffer Recognition (ABR)
0.1°C
Offset in 0.1°C Increments; Offset Range: ±5°C
BNC, 2.5 mm Phono Plug, Reference Pin Tip, Ground
Up to ±150 mV
AC/DC 9 V Adapter (110/220 VAC, 50/60 Hz)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ accumet™ FE150 Benchtop pH Meter > FE150 Benchtop pH Meter; Includes: Meter, Electrode Stand, Universal Power Adapter (90 to 240 V, 50/60 Hz); Type: Standard Set; Calibration Points: 1 to 5 with Automatic Buffer Recognition (ABR)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.