missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ XL250 Benchtop pH/ISE Dual Channel Meters

Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

1480.00 € - 2080.00 €

Specifications

Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Accuracy Multiple
Accuracy (ISE) 0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
View More Specs

Products 3
Product Code Brand Model Includes Price Quantity & Availability  
Product Code Brand Model Includes Price Quantity & Availability  
12850643
View Documents
Fisherbrand™
01X605301
XL250 Dual pH/ISE Meter Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply
1480.00 €
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
12860643
View Documents
Fisherbrand™
01X610101
XL250 Dual pH/ISE Kit Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply
1610.00 €
Each
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
12890643
View Documents
Fisherbrand™
01X610301
XL250 Dual pH/ISE Ammonia Kit Meter, 13-620-509 ammonia electrode, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply
2080.00 €
Each
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
Description

Description

Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display two pH or ISE measurements simultaneously with dual channel BNC inputs
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ISE Calibration Points: 2 to 6 points
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: Two BNC, 2 mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Includes:
13-636-XL250A: accumet XL250 pH/ISE dual chan. meter, electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250F: accumet XL250 pH/ISE dual chan. meter, fluoride electrode (13-620-629), TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250N: accumet XL250 pH/ISE dual chan. meter, ammonia electrode (13-620-509), TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

13-636-XL250: accumet XL250 pH/ISE dual chan. meter, TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply.

XL250ACERT: accumet XL250 pH/ISE dual chan. meter, electrode stand, RS232 and USB cables, 100/240V univ power supply, NIST certif.

XL250CERT: accumet XL250 pH/ISE dual chan. meter, TRIS-compat double junc refillable pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V univ power supply, NIST certif.

Certifications

CE, TUV 3-1, FCC Class A

Warranty

3 years on meter

Specifications

Specifications

RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
0.5% full scale (monovalent ion), 1% full scale (divalent ion)
±0.002
Up to 6 preset or 5 custom
3 to 86400 sec.
Multiple
±2000 mV
2/3/4 Digits
2 BNC, 2 ATC
Multiple
±0.3°C
3 Years
9.68 in.
17.3 cm
9.9 cm
1.13 kg
50/60Hz
Multiple
±0.2 mV
2 to 6
NIST, ISO, FDA, USDA, EPA
2000
LCD
-2 to 20
0.1/0.01/0.001
±2000 mV
0.1mV
0.1°C
24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
24.6 cm
3.89 in.
2.5 lb.
100/240V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.