missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuPrime™ SuperMix I

AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.

Brand:  Invitrogen™ 12342010

 View more versions of this product

Product Code. 10564233

  • 552.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix I sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.

In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.

AccuPrime SuperMix I is designed for amplification of genomic DNA amplicons (> 200 bp), plasmid DNA, or cDNA templates. Alternatively, AccuPrime SuperMix II is designed to amplify a broader range of amplicon sizes up to 4 kb.

• Built-in hot start

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

AccuPrime SuperMix I is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 200 amplification reactions of 25 μL each are provided. AccuPrime SuperMix I Components: 40 mM Tris-HCl (pH 8.4), 100 mM KCl, 3 mM MgCl2, 400 μM dGTP, 400 μM dATP, 400 μM dTTP, 400 μM dCTP, AccuPrime Taq DNA Polymerase, thermostable AccuPrime protein, stabilizers.

200 Reactions: 2 x 1.25 mL.
Store at -20°C in a non-frost-free freezer.

Hot-start PCR, High-fidelity PCR
200 Reactions
Low
2X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.