Learn More
Draeger Safety Accuro™ Gas Detector Pump
Draeger Safety Accuro™ Gas Detector Pump is a manually operated bellows pump that draws a calibrated 100 mL sample through the short-term Draeger-Tubes.
Brand:  Draeger Safety 6400260
Product Code. 10693781
  • 1530.00 € / Each
Explore more special offers
Description
Description
  • One-Hand Operation
  • Very Low Maintenance
  • End-of-Stroke Indicator
  • Built-in automatic Stroke Counter
Specifications
Specifications
205 x 240 x 60 mm
0.92 kg