Learn More
Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v2.2, seat license
AccuSEQ Real-Time PCR Detection Software v2.2 is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ and ViralSEQ assays.
Brand:  Applied Biosystems™ A58644
Product Code. 18723644
  • 3920.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
7500 Fast System
Real-Time PCR Software
Real Time PCR (qPCR)
1 license
AccuSEQ™
Real Time PCR Detection Software
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ AccuSEQ™ Real-Time PCR Detection Software v2.2, seat license > 1 License