missing translation for 'onlineSavingsMsg'
Learn More

Ansell AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves

Provides optimal solutions for controlled and sterile environments, respectively. Ansell Edmont™ AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves are made with a specific natural rubber formulation that offers a high level of comfortable, sensitive, precise handling.

490.00 €

Specifications

Cleanroom Class Class 100 (ISO 5)
Color Natural
Latex Free No
Material Latex
Product Type Cleanroom Gloves
View More Specs

Products 7
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15265183
View Documents
Ansell
91325060
6 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11745773
View Documents
Ansell
91325065
6.5 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11755773
View Documents
Ansell
91325075
7.5 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11776713
View Documents
Ansell
91325070
7 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11786713
View Documents
Ansell
91325080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15275183
View Documents
Ansell
91325090
9 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11765773
View Documents
Ansell
91325085
8.5 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • With textured fingertips
  • Excellent technical properties
  • Low allergenic content minimizes the risk of latex-associated allergies for the wearer
  • Ideal for biotechnologies, medical device manufacturing, laboratory analysis, pharmaceuticals and the flat panel industry
Specifications

Specifications

Class 100 (ISO 5)
No
Cleanroom Gloves
Natural
200 Pairs
Natural
Latex
9.8 mm
Straight
91-325
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.