missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Product Code. 10473585
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58052-80-5
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00149559
InChI Key MZSSRMMSFLVKPK-UHFFFAOYSA-Q
Synonym 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
PubChem CID 2723814
SMILES CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 97.0°C
Color Beige
Boiling Point 110.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 g
Beilstein 26, I, 3
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane;trihydrate
Formula Weight 183.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde ammonia trimer, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Acetaldehyde ammonia trimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'