Learn More
Acetaldehyde dimethyl acetal, 98+%, Thermo Scientific Chemicals
35.98 € - 369.84 €
Chemical Identifiers
CAS | 534-15-6 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.122 |
MDL Number | MFCD00008493 |
InChI Key | SPEUIVXLLWOEMJ-UHFFFAOYSA-N |
Synonym | dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane |
PubChem CID | 10795 |
IUPAC Name | 1,1-dimethoxyethane |
SMILES | CC(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494343
|
Thermo Scientific Alfa Aesar
A10939.14 |
25 g |
35.98 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404353
|
Thermo Scientific Alfa Aesar
A10939.22 |
100 g |
95.58 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414353
|
Thermo Scientific Alfa Aesar
A10939.36 |
500 g |
369.84 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.
Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
534-15-6 | |
90.122 | |
SPEUIVXLLWOEMJ-UHFFFAOYSA-N | |
10795 | |
CC(OC)OC |
C4H10O2 | |
MFCD00008493 | |
dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane | |
1,1-dimethoxyethane |
Specifications
534-15-6 | |
0.852 | |
−17°C (1°F) | |
C4H10O2 | |
MFCD00008493 | |
1697039 | |
14,3226 | |
Soluble in water (1000mg/ml at 25°C),alcohols,chloroform,ether,and acetone. | |
CC(OC)OC | |
90.122 | |
90.12 | |
Acetaldehyde dimethyl acetal |
-113°C | |
63°C to 65°C | |
Sweet | |
1.367 | |
UN2377 | |
Moisture sensitive | |
dimethyl acetal, acetaldehyde dimethyl acetal, ethane, 1,1-dimethoxy, dimethylacetal, dimethyl aldehyde, ethylidene dimethyl ether, methyl formyl, acetaldehyde, dimethyl acetal, acetaldehyde methyl acetal, 3-methyl-2,4-dioxapentane | |
SPEUIVXLLWOEMJ-UHFFFAOYSA-N | |
1,1-dimethoxyethane | |
10795 | |
≥98% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,1-DIMETHOXYETHANE
missing translation for 'einecsNumber' : 208-589-8
missing translation for 'rtecsNumber' : AB2825000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Acetaldehyde dimethyl acetal
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : 1,1-Dimethoxyethane
RUO – Research Use Only