missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, syn + anti, 98%

Product Code. 11413374
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11413374

Brand: Thermo Scientific Alfa Aesar undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-29-9 | C2H5NO | 59.07 g/mol

Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

Solubility
Soluble in water, ethanol and ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (NZ)-N-ethylidenehydroxylamine
SMILES C\C=N\O

Specifications

Density 0.969
Boiling Point 115°C to 117°C
Flash Point 36°C (96°F)
Odor Characteristic
Refractive Index 1.426
Quantity 250 g
UN Number UN2332
Beilstein 1209252
Merck Index 14,42
Solubility Information Soluble in water,ethanol and ether.
Formula Weight 59.07
Percent Purity 98%
Chemical Name or Material Acetaldoxime, syn + anti
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.