missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
125.00 € - 176.00 €
Chemical Identifiers
CAS | 6131-90-4, 7732-18-5 |
---|---|
Molecular Formula | C2H9NaO5 |
Molecular Weight (g/mol) | 136.079 |
InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
PubChem CID | 23665404 |
ChEBI | CHEBI:32138 |
IUPAC Name | sodium;acetate;trihydrate |
SMILES | CC(=O)[O-].O.O.O.[Na+] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11496078
|
SPEX Certiprep
5272 |
125 mL | Triple-leached LDPE bottle | N/A | |||||
15255796
|
SPEX Certiprep
5272 |
125 mL | Triple-leached LDPE bottle |
167.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12775215
|
SPEX Certiprep
AS-ACE9-2X |
500 mL | Triple-leached LDPE bottle |
125.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15245796
|
SPEX Certiprep
5262 |
500 mL | Triple-leached LDPE bottle |
174.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11486078
|
SPEX Certiprep
5262 |
500 mL | Triple-leached LDPE bottle | N/A | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
CH3COO- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
6131-90-4, 7732-18-5 | |
136.079 | |
23665404 | |
sodium;acetate;trihydrate |
C2H9NaO5 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
Specifications
6131-90-4, 7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
C2H9NaO5 | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
Sodium Acetate,distilled water,conductivity or of similar purity | |
Acetate Standard, For Ion Chromatography |
0.1, 99.9 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
1000mg/L | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only