Learn More
Acetic acid, 4% v/v aq. soln., Thermo Scientific Chemicals
22.70 € - 50.45 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445020
|
Thermo Scientific Alfa Aesar
035572.AP |
500 mL | N/A | ||||||
13552310
|
Thermo Scientific Alfa Aesar
035572.AP |
500 mL |
28.74 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373667
|
Thermo Scientific Alfa Aesar
035572.K2 |
1 L | N/A | ||||||
13562310
|
Thermo Scientific Alfa Aesar
035572.K2 |
1 L |
50.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
4% v/v aq. soln. | |
60.05 | |
CHEBI:15366 | |
Pungent | |
Acetic acid |
C2H4O2 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
acetic acid | |
176 | |
60.05 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 200-580-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.