Learn More
Acetic acid, 99.7+%, ACS reagent, Thermo Scientific Chemicals
37.60 € - 301.00 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10005920
|
Thermo Scientific Acros
423225000 |
500 mL | Glass Bottle |
37.60 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005910
|
Thermo Scientific Acros
423220025 |
2.5 L | Glass Bottle |
99.30 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358483
|
Thermo Scientific Acros
423220100 |
10 L | Plastic drum |
301.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
100.0 | |
C2H4O2 | |
MFCD00036152 | |
02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.70% | |
1.0480g/mL | |
0.001% max. | |
Acetic Acid |
99.7 | |
99.7+% | |
CH3COOH | |
02, 96 | |
15, 54 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
ACS Reagent | |
Liquid | |
1.52kPa at 20°C |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
- Acetic acid
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only