missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 99.8%, for biochemistry, Thermo Scientific Chemicals

46.70 € - 226.00 €

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10677112
View Documents Promotion Details
Thermo Scientific Acros
148930010
1 L Glass Bottle
46.70 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10336600
View Documents Promotion Details
Thermo Scientific Acros
148930025
2.5 L Glass Bottle
90.50 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10721974
View Documents Promotion Details
Thermo Scientific Acros
148930100
10 L Plastic drum This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

(neat), (neat) at 0nm, 0.02 max. at 280nm, 0.1 max. at 260nm
16.0°C to 16.5°C
99.75
99.8%
CH3COOH
2789
02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Biochemical
40°C
Liquid
1.048
Acetic Acid, For biochemistry
64-19-7
117.0°C to 118.0°C
100.0
C2H4O2
MFCD00036152
02, 96
15, 54
Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
CC(O)=O
60.05
CHEBI:15366
99.8%
1.0480g/mL
Authentic
1.3700 to 1.3730
1.52kPa at 20°C
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

Product Identifier
  • Acetic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Acetic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only