missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, eluent additive for LC-MS, Honeywell™

Product Code. 15625660
Click to view available options
Quantity:
50 mL

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 64-19-7
Color Colorless
Molecular Formula C2H4O2
Linear Formula CH3CO2H
MDL Number MFCD00036152
UN Number UN2789
Beilstein 506007
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05g/mol
Density 1.049g/mL at 25°C
Quantity 50 mL
Explosion Limit 4% at 59°F, 16% at 92°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid, eluent additive for LC-MS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'