missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Environmental Grade Plus, 99.4% min, Thermo Scientific™
Click to view available options
Quantity:
250 mL
500 mL
Dimensione della confezione:
250mL
500mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| pH | 2.5 |
| Melting Point | 16.6°C |
| Boiling Point | 118.1°C |
| UN Number | UN2789 |
| Merck Index | 14,55 |
| Formula Weight | 60.05 |
| Percent Purity | ≤99.4% |
| Grade | Environmental Plus |
| Density | 1.049g/mL |
| Quantity | 250 mL |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- Acetic acid, glacial
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto