missing translation for 'onlineSavingsMsg'
Learn More
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals Product Code.: 11475160

Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals

Product Code. 11475160
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2 L
4 x 500 mL
This item is not returnable. View return policy

Product Code. 11475160

Brand: Thermo Scientific Alfa Aesar 036289.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.6°C
Density 1.049
Boiling Point 118.1°C
Flash Point 40°C (104°F)
Assay Percent Range 99.7+%
Refractive Index 1.3721
Quantity 500 mL
UN Number 2789
Merck Index 14,55
Solubility Information Miscible with water.
Formula Weight 60.05
Percent Purity ≥99.7%
Grade ACS
Physical Form Liquid
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN03L023 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number036289.AE,036289.K3,036289.AY,036289.AP
Lot NumberM28L042 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number036289.AE,036289.K3,036289.AY,036289.AP
Lot NumberM22L008 Certificate TypeCertificate of Analysis Date24/01/2025 Catalog Number036289.AE,036289.AY,036289.K3,036289.AP
Lot NumberM13L019 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number036289.AE,036289.AY,036289.K3,036289.AP
Lot NumberM13L014 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number036289.AE,036289.AY,036289.K3,036289.AP
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, glacial
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.