missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial Min. 99%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
76.40 € - 145.00 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375890
|
Fisher Chemical
A507-P500 |
500 mL |
76.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395890
|
Fisher Chemical
A507-P1 |
1 L |
98.25 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11315900
|
Fisher Chemical
A507-P212 |
2.5 L |
145.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
CC(O)=O |
C2H4O2 | |
MFCD00036152 | |
acetic acid |
Specifications
2.5 | |
16°C | |
Colorless | |
MFCD00036152 | |
CC(O)=O | |
60.05 | |
Liquid | |
1.52kPa at 20°C |
64-19-7 | |
117°C | |
C2H4O2 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
60.05g/mol | |
2.1 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Acetic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title