missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial (Optima™), Fisher Chemical™
364.00 € - 675.00 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11994891
|
Fisher Chemical
A465-250 |
250 mL |
364.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11904901
|
Fisher Chemical
A465-500 |
500 mL | FEP Bottle |
367.72 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984891
|
Fisher Chemical
A465-1 |
1 L |
675.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Certificate of Analysis included with each bottle.
- Ideal for analysis by ICP-MS and high-resolution ICP-MS
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
CC(O)=O |
C2H4O2 | |
MFCD00036152 | |
acetic acid |
Specifications
2.5 | |
16°C | |
Colorless | |
MFCD00036152 | |
CC(O)=O | |
60.05 | |
Liquid | |
1.52kPa at 20°C |
64-19-7 | |
117°C | |
C2H4O2 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
60.05g/mol | |
2.1 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Flammable liquid Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title