missing translation for 'onlineSavingsMsg'
Learn More
Acetic Acid Glacial, Pure, for Non Aqueous Titrations, Fisher Chemical Product Code.: 10375020

Acetic Acid Glacial, Pure, for Non Aqueous Titrations, Fisher Chemical

Product Code. 10375020
2.5L, Plastic Coated Glass bottle
Click to view available options
2.5L, Plastic Coated Glass bottle
This item is not returnable. View return policy

Product Code. 10375020

Brand: Fisher Chemical A/0420/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH 2.5
CAS 64-19-7
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Molecular Formula C2H4O2
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
Formula Weight 60.05g/mol
Quantity 2.5 L
Packaging Plastic Coated Glass bottle
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2523660 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberA/0420/PB17
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic Acid Glacial, Pure, for Non Aqueous Titrations, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.