missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial, for Trace Metal Analysis, Primar Plus™, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Specifications
Specifications
pH | 2.5 |
Melting Point | 16°C |
Boiling Point | 117°C |
Color | Colorless |
Formula Weight | 60.05g/mol |
Quantity | 500 mL |
Packaging | HDPE plastic bottle |
Physical Form | Liquid |
Vapor Density | 2.1 |
Vapor Pressure | 1.52kPa at 20°C |
Safety and Handling
Product Identifier
- Acetic Acid
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'