missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
48.50 € - 248.00 €
Chemical Identifiers
CAS | 127-08-2 |
---|---|
Molecular Formula | C2H3KO2 |
Molecular Weight (g/mol) | 98.142 |
MDL Number | MFCD00012458 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
IUPAC Name | potassium;acetate |
SMILES | CC(=O)[O-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10338640
|
Thermo Scientific Acros
217105000 |
500 g | Plastic bottle |
48.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10072830
|
Thermo Scientific Acros
217100010 |
1 kg | Plastic bottle |
82.20 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177303
|
Thermo Scientific Acros
217100050 |
5 kg | Plastic drum |
248.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
127-08-2 | |
98.142 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
Specifications
7.5 to 8.5 (5% soln. at 20°C) | |
292.0°C | |
100.0 | |
99+% | |
CH3CO2K | |
02, 96 | |
15, 7726 | |
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
≥99% | |
>250°C | |
Granular Powder |
127-08-2 | |
99.0 | |
White | |
C2H3KO2 | |
MFCD00012458 | |
01,906; 06,475 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Analytical | |
Authentic | |
Acetic acid, potassium salt, For analysis ACS |
RUO – Research Use Only