missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

65.11 € - 2151.68 €

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 12
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15643920
View Documents Promotion Details
Honeywell-Fluka
33209-1L-GL
1 L Glass Bottle
65.11 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15633920
View Documents Promotion Details
Honeywell-Fluka
33209-1L
1 L Plastic Bottle
65.11 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15663920
View Documents Promotion Details
Honeywell-Fluka
33209-2.5L-GL
2.5 L Glass Bottle
142.99 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15653920
View Documents Promotion Details
Honeywell-Fluka
33209-2.5L
2.5 L Plastic Bottle
140.09 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15613930
View Documents Promotion Details
Honeywell-Fluka
33209-5L
5 L Plastic Bottle
293.07 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15643930
View Documents Promotion Details
Honeywell-Fluka
33209-6X1L-GL
6 x 1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15633930
View Documents Promotion Details
Honeywell-Fluka
33209-6X1L
6 x 1 L Plastic Bottle
338.55 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15603930
View Documents Promotion Details
Honeywell-Fluka
33209-4X2.5L-GL
4 x 2.5 L Glass Bottle
536.02 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15693920
View Documents Promotion Details
Honeywell-Fluka
33209-4X2.5L
4 x 2.5 L Plastic Bottle
487.29 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15683920
View Documents Promotion Details
Honeywell-Fluka
33209-30KG
30 kg Plastic Pail
269.17 €
30kg
Please to purchase this item. Need a web account? Register with us today!
15623930
View Documents Promotion Details
Honeywell-Fluka
33209-60KG
60 kg Plastic Pail
509.78 €
60kg
Please to purchase this item. Need a web account? Register with us today!
15673920
View Documents Promotion Details
Honeywell-Fluka
33209-200L
200 L Poly Drum
2151.68 €
200 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Spezifikation

64-19-7
CH3CO2H
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
≥99.8%
1.049g/mL at 25°C
40°C (104°F)
2.07 (vs air)
C2H4O2
MFCD00036152
506007
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05g/mol
ACS/puriss. p.a.
16% at 92°F, 4% at 59°F
800°F
Acetic acid
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378

missing translation for 'rtecsNumber' : AF1225000

Produktidentifikator
  • Acetic acid

Signalwort
  • Danger

Gefahrenkategorie
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Sicherheitshinweis
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate