missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, ReagentPlus™, Honeywell
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Beilstein | 506007 |
Formula Weight | 60.05g/mol |
Percent Purity | ≥99% |
Grade | ReagentPlus™ |
Density | 1.049g/mL at 25°C |
Quantity | 100 mL |
Explosion Limit | 16% at 92°F, 4% at 59°F |
Flash Point | 40°C (104°F) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'