Learn More
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
37.29 € - 113.57 €
Chemical Identifiers
CAS | 127-09-3, 6131-90-4 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
IUPAC Name | sodium acetate |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206800
|
Thermo Scientific Acros
217115000 |
500 g | Plastic bottle |
37.29 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10462791
|
Thermo Scientific Acros
217110010 |
1 kg | Plastic bottle |
56.48 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236750
|
Thermo Scientific Acros
217110025 |
2.5 kg | Plastic bottle |
113.57 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
127-09-3, 6131-90-4 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
sodium acetate |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 | |
[Na+].CC([O-])=O |
Specifications
7.5 to 9.0 (5 % at 20°C) | |
58.0°C | |
100.0 | |
99.5% | |
CH3COONa·3H2O | |
02, 96 | |
15, 8709 | |
Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
99.5% | |
Authentic | |
Acetic acid, sodium salt trihydrate, p.a. |
127-09-3, 6131-90-4 | |
99.4 | |
White | |
C2H3NaO2 | |
MFCD00012459 | |
01,1024; 05,591 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate | |
517045 | |
136.08 | |
Analytical | |
Crystalline |
Safety and Handling
HYGROSCOPIC
missing translation for 'rtecsNumber' : AJ4580000
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.