missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals
54.67 € - 188.94 €
Chemical Identifiers
CAS | 127-09-3, 6131-90-4 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011140
|
Thermo Scientific Acros
123240010 |
1 kg | Plastic Bottle |
54.67 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687963
|
Thermo Scientific Acros
123240050 |
5 kg | Plastic drum |
188.94 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-09-3, 6131-90-4 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Specifications
127-09-3, 6131-90-4 | |
99.0 | |
White | |
C2H3NaO2 | |
MFCD00012459 | |
01,1024; 05,591 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate | |
517045 | |
136.08 | |
Extra Pure | |
Adhering Crystals |
58.0°C | |
100.0 | |
99+% | |
CH3COONa·3H2O | |
02, 96 | |
15, 8709 | |
Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99% | |
Authentic | |
Acetic acid, sodium salt trihydrate |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals