missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid solution, 1.0M, Honeywell Fluka™
Click to view available options
Quantity:
1 L
10 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Linear Formula | CH3CO2H |
UN Number | NONH for all modes of transport |
Beilstein | 506007 |
Concentration | 1.0M |
Formula Weight | 60.05g/mol |
Quantity | 10 L |
Chemical Name or Material | Acetic acid solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction