missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TraceSELECT™ Ultra, Honeywell Fluka™

Product Code. 15670870
Click to view available options
Quantity:
250 mL
500 mL
1 L

TraceSELECT™ Ultra, for ultratrace analysis,≥99.0%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Linear Formula CH3CO2H
UN Number 2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99%
Grade TraceSELECT™ Ultra
Density 1.049g/mL at 25°C
Quantity 500 mL
Explosion Limit 4% (59°F), 16% (92°F)
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Plastic Bottle
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'