missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid – Triethylamine Solution 1:1, Honeywell Fluka™

Product Code. 15660900
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

for HPLC, 2M:2M concentrate, in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5204-74-0
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00151725
InChI Key AVBGNFCMKJOFIN-UHFFFAOYSA-N
Synonym triethylammonium acetate, teaa, acetic acid, compd. with n,n-diethylethanamine 1:1, acetic acid; n,n-diethylethanamine, ethanamine, n,n-diethyl-, acetate, triethylamine acetate, hoac et3n, acetic acid triethylamine, acetic acid; triethylamine, triethylammonium acetate solution
PubChem CID 165295
IUPAC Name acetic acid;N,N-diethylethanamine
SMILES CCN(CC)CC.CC(=O)O

Specifications

Linear Formula CH3COOH · N(CH2CH3)3
UN Number NONH for all modes of transport
Beilstein 3688333
Formula Weight 161.24g/mol
Density 1.010g/mL at 20°C
Quantity 100 mL
Chemical Name or Material Acetic Acid – Triethylamine Solution 1:1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'