Learn More
Acetic Anhydride, Reagent ACS, +97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.087 g/mL |
Assay Percent Range | 97+% |
Packaging | Glass bottle |
Linear Formula | (CH3CO)2O |
Quantity | 2.5 L |
Beilstein | 02, 166 |
Residue after Evaporation | 0.003% max. |
Fieser | 01,3; 02,7; 05,3; 06,1; 07,1; 08,1; 09,1; 11,1; 12,1; 13,93; 14,150; 15,1 |
Merck Index | 15, 55 |
SMILES | CC(=O)OC(C)=O |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466690 | Certificate of Analysis | 10/10/2024 | 423230010,423235000,423230025 |
A0464549 | Certificate of Analysis | 18/07/2024 | 423230010,423235000,423230025 |
A0463041 | Certificate of Analysis | 23/05/2024 | 423230010,423235000,423230025 |
A0458762 | Certificate of Analysis | 14/12/2023 | 423230010,423230050,423235000,423230025 |
B0542556B | Certificate of Analysis | 14/11/2023 | 423230010,423230050,423235000,423230025 |
Safety and Handling



- Acetic anhydride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.