missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ≥99.9% (GC), Honeywell™ Fluka™ TraceSELECT™

Product Code. 15676720
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

TraceSELECT™, for inorganic trace analysis,≥99.9%

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Color Colorless
Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Molecular Formula C3H6O
Linear Formula CH3COCH3
MDL Number MFCD00008765
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity ≥99.9% (GC)
Grade TraceSELECT™
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acetone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'