missing translation for 'onlineSavingsMsg'
Learn More
Acetone, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10225900

Acetone, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10225900
2.5 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10225900

Brand: Thermo Scientific Acros 177170025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
CAS Min % 99.0
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Boiling Point 56.0°C
Quantity 2.5 L
Flash Point -20°C (-4°C)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.79
Linear Formula CH3COCH3
Vapor Pressure Vapor Pressure: 247mbar at 20°C
UN Number 1090; Colorless
Viscosity 0.32 mPa.s (20°C)
Beilstein 01, 635
Water 0.5% max. (K.F.)
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Acetone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2522615 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177170025,177170100,177170050,177170010,177170250
Lot Number2421228 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177170025,177170100,177170050,177170010,177170250
Lot Number2419153 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177170025,177170100,177170050,177170010,177170250
Lot Number2524360 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177170025,177170100,177170050,177170010,177170250
Lot Number2405502 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177170025,177170100,177170050,177170010,177170250
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.