missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
2 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2 L
2.5 L
20 L
500mL
Pack of 4
Pack of 6
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -94°C (literature) |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Density | 0.791 g/mL at 25 °C(lit.) |
| Boiling Point | 55°C to 57°C |
| Quantity | 2 L |
| Flash Point | −17°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3COCH3 |
| UN Number | UN1090 |
| Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction