missing translation for 'onlineSavingsMsg'
Learn More

Acetone, BAKER ANALYZED Pesticide Reagent, J.T. Baker™

Product Code. 15253388
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Specially developed for use in evaluation of pesticide and insecticide residues in food, feed, water or soil samples. Acetone, BAKER ANALYZED Pesticide Reagent, J.T. Baker™ is suitable for all analysis of common pesticides.

J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:

  • Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
  • Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
  • Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation

Specifications

pH 5 to 6 (at 20°C)
Melting Point -94.9°C
CAS 67-64-1
Color Colorless
Acidity Max. 0.0005meq/g
Density 0.79g/cm3 at 20°C
Boiling Point 55.8°C to 56.6°C
Quantity 2.5 L
Explosion Limit 2.6 - 13 Vol %
Flash Point −20°C
Ignition Point 465°C
Odor Fruit-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula C3H6O
MDL Number MFCD00008765
Viscosity 32 mPas at 25°C
Water Max. 0.3%
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Fully miscible in water
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity Min. 99.8%
Grade BAKER ANALYZED™ Pesticide
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.