missing translation for 'onlineSavingsMsg'
Learn More
Acetone, N.F., A.C.S., J.T. Baker™ Product Code.: 15588004

Acetone, N.F., A.C.S., J.T. Baker™

Product Code. 15588004
4 L, Poly Bottle
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15588004

Brand: Avantor J.T.Baker 246203

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 67-64-1
Quantity 4 L
Flash Point −20.00°C
Packaging Poly Bottle
Specific Gravity 1L = 0.79kg
Molecular Formula C3H6O
MDL Number MFCD00008765
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08
Grade ACS/NF
Chemical Name or Material Acetone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, N.F., A.C.S., J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.